p-Toluenesulfonic acid, sodium salt
p-Toluenesulfonic acid, sodium salt
p-Toluenesulfonic acid, sodium salt
Thermo Scientific Chemicals

p-Toluenesulfonic acid, sodium salt

CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 421235000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS657-84-1
IUPAC Namesodium 4-methylbenzene-1-sulfonate
Molecular FormulaC7H7NaO3S
InChI KeyKVCGISUBCHHTDD-UHFFFAOYSA-M
SMILES[Na+].CC1=CC=C(C=C1)S([O-])(=O)=O
さらに表示
Appearance (Color)White to off-white
Appearance (Form)Powder
Titration after Ion exchange>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only