Benzyl mercaptan, 99%
Benzyl mercaptan, 99%
Benzyl mercaptan, 99%
Acros Organics

Benzyl mercaptan, 99%

CAS: 100-53-8 | C7H8S | 124.20 g/mol
Have Questions?
Catalog NumberQuantityPackaging
421265000
also known as 42126
500 gGlass bottle
Catalog number 421265000
also known as 42126
HSN 29309099
Price (INR)
51,761.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
51,761.00
Each
Chemical Identifiers
CAS100-53-8
IUPAC Namephenylmethanethiol
Molecular FormulaC7H8S
InChI KeyUENWRTRMUIOCKN-UHFFFAOYSA-N
SMILESSCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only