Trichloroisocyanuric acid, 99%
Trichloroisocyanuric acid, 99%
Trichloroisocyanuric acid, 99%
Thermo Scientific Chemicals

Trichloroisocyanuric acid, 99%

CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 421540050
Price (EUR)
354,00
Each
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
354,00
Each
Chemical Identifiers
CAS87-90-1
IUPAC Nametrichloro-1,3,5-triazinane-2,4,6-trione
Molecular FormulaC3Cl3N3O3
InChI KeyYRIZYWQGELRKNT-UHFFFAOYSA-N
SMILESClN1C(=O)N(Cl)C(=O)N(Cl)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
Titration Iodometric>=98.5 %
Appearance (Form)Powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only