Tridodecylamine, 90%
Tridodecylamine, 90%
Tridodecylamine, 90%
Thermo Scientific Chemicals

Tridodecylamine, 90%

CAS: 102-87-4 | C36H75N | 522 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 421621000
Price (EUR)
190,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
190,00
Each
Chemical Identifiers
CAS102-87-4
IUPAC Nametridodecylamine
Molecular FormulaC36H75N
InChI KeySWZDQOUHBYYPJD-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=88 %
Appearance (Form)Liquid or low melting solid
Appearance (Color)Clear colorless to light yellow
Refractive index1.4550 to 1.4610 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only