Triethanolamine, 99+%, Thermo Scientific Chemicals
Triethanolamine, 99+%, Thermo Scientific Chemicals
Triethanolamine, 99+%, Thermo Scientific Chemicals
Triethanolamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethanolamine, 99+%, Thermo Scientific Chemicals

Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 421630025
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS102-71-6
IUPAC Name2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC6H15NO3
InChI KeyGSEJCLTVZPLZKY-UHFFFAOYSA-N
SMILESOCCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Viscous liquid
Refractive index1.4830 to 1.4860 (20°C, 589 nm)
Titration with HClO4>=99.0 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only