Trilaurin, 98%
Trilaurin, 98%
Trilaurin, 98%
Thermo Scientific Chemicals

Trilaurin, 98%

CAS: 538-24-9 | C39H74O6 | 639.01 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 421830250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS538-24-9
IUPAC Name1,3-bis(dodecanoyloxy)propan-2-yl dodecanoate
Molecular FormulaC39H74O6
InChI KeyVMPHSYLJUKZBJJ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
TLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only