You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Gel Electrophoresis Equipment and Supplies
▸
Gel Electrophoresis Reagents and Buffers
▸
Gel Electrophoresis Buffers and Diluents
▸
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid, ≥98%, purified, GMP, J.T.Baker™
Avantor™
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid, ≥98%, purified, GMP, J.T.Baker™
GMP Manufactured Product.
Have Questions?
Change view
Catalog Number
Quantity
4220-08
25 kg
4220-06
10 kg
2 Options
Catalog number
4220-08
Price (CLP)
-
Request A Quote
Quantity:
25 kg
GMP Manufactured Product.
Specifications
Purity or Quality Grade
Purified
Quantity
25 kg
Form
Crystalline Powder
Unit Size
Each