2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
Thermo Scientific Chemicals

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%

CAS: 6846-50-0 | C16H30O4 | 286.4 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
1 kg
Packaging:
Glass bottle
產品號碼 422000010
價格 (TWD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS6846-50-0
IUPAC Name2,2,4-trimethyl-1-[(2-methylpropanoyl)oxy]pentan-3-yl 2-methylpropanoate
Molecular FormulaC16H30O4
InChI KeyOMVSWZDEEGIJJI-UHFFFAOYNA-N
SMILESCC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only