2,4,6-Triphenyl-s-triazine, 98+%
2,4,6-Triphenyl-s-triazine, 98+%
2,4,6-Triphenyl-s-triazine, 98+%
Thermo Scientific Chemicals

2,4,6-Triphenyl-s-triazine, 98+%

CAS: 493-77-6 | C21H15N3 | 309.37 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 422190250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS493-77-6
IUPAC Nametriphenyl-1,3,5-triazine
Molecular FormulaC21H15N3
InChI KeyHBQUOLGAXBYZGR-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Appearance (Color)White to almost white
HPLC>=98.0 %
Appearance (Form)Crystalline powder
Melting point231°C to 236°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only