Valeramide, 97%
Valeramide, 97%
Valeramide, 97%
Thermo Scientific Chemicals

Valeramide, 97%

CAS: 626-97-1 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 422440025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS626-97-1
IUPAC Namepentanamide
Molecular FormulaC5H11NO
InChI KeyIPWFJLQDVFKJDU-UHFFFAOYSA-N
SMILESCCCCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point101°C to 106°C
Appearance (Color)White
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Glistening powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only