N-(4-Vinylbenzyl)-N,N-dimethylamine, 90%, technical, stabilized
N-(4-Vinylbenzyl)-N,N-dimethylamine, 90%, technical, stabilized
N-(4-Vinylbenzyl)-N,N-dimethylamine, 90%, technical, stabilized
Thermo Scientific Chemicals

N-(4-Vinylbenzyl)-N,N-dimethylamine, 90%, technical, stabilized

CAS: 2245-52-5 | C11H16N | 162.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 422550050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2245-52-5
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Infrared spectrumConforms
Refractive index1.5360 to 1.5400 (20°C, 589 nm)
Stabilizer0.1 % Hydroquinone
Appearance (Color)Clear yellow to orange to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only