Vinylbenzyl trimethylammonium chloride, 97%
Vinylbenzyl trimethylammonium chloride, 97%
Vinylbenzyl trimethylammonium chloride, 97%
Vinylbenzyl trimethylammonium chloride, 97%
Thermo Scientific Chemicals

Vinylbenzyl trimethylammonium chloride, 97%

CAS: 26616-35-3 | C12H18ClN | 211.73 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 422560250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26616-35-3
Molecular FormulaC12H20ClN
SMILES[Cl-].*-C=C.C[N+](C)(C)C-*.C1=CC=CC=C1
Molecular Weight (g/mol)213.75
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only