Vitamin D2, 97+%
Vitamin D2, 97+%
Vitamin D2, 97+%
Thermo Scientific Chemicals

Vitamin D2, 97+%

Vitamin D2, or Ergocalciferol, CAS # 50-14-6, a fat-soluble hormone that is important for calcium and phosphorus absorption and metabolism. | CAS: 50-14-6 | C28H44O | 396.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 422630050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS50-14-6
IUPAC Name(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
Molecular FormulaC28H44O
InChI KeyMECHNRXZTMCUDQ-VLOQVYPSSA-N
SMILESCC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Melting point113°C to 119°C
HPLC97.0 to 103.0 %
Specific optical rotation+100° to +106° (20°C, 589 nm) (c=1,5; EtOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vitamin D2 (ergocalciferol) is a form of vitamin D, a fat-soluble hormone found in food sources
  • Vitamin D is important for calcium and phosphorus homeostasis, and can play a role in many biological processes including cell growth
  • Vitamin D2 has a lower binding affinity than Vitamin D3 for vitamin D binding protein

Applications

  • Research studies show that Vitamin D2 treatment in rats inhibits bladder tumor promotion
  • Studies show that the treatment of lung carcinoma cells with vitamin D2 in vitro resulted in increased ACE2 protein levels
  • Vitamin D2 has been seen to interfere with prion protein oligomerization in vitro
RUO – Research Use Only

General References:

  1. Yazawa, Y., et al., 2000. Antitumor promoting effect of an active component of Polyporus, ergosterol and related compounds on rat urinary bladder carcinogenesis in a short-term test with concanavalin A. Biological & pharmaceutical bulletin. 23(11): 1298-302.
  2. Bártová, E.; Legartová, S.; Krejčí, J.; Arcidiacono, O.A. Cell differentiation and aging accompanied by depletion of the ACE2 protein. Aging (Albany NY). 2020, 12(22), 22495-22508.
  3. Suenaga, M.; Hiramoto, Y.; Matsunaga, Y. Vitamin D 2 interacts with Human PrP(c) (90-231) and breaks PrP(c) oligomerization in vitro. Prion. 2013, 7(4), 312-8.
  4. Soto-Dávila, M.; Valderrama, K.; Inkpen, S.M.; Hall, J.R.; Rise, M.L.; Santander, J. Effects of Vitamin D2 (Ergocalciferol) and D3 (Cholecalciferol) on Atlantic Salmon (Salmo salar) Primary Macrophage Immune Response to Aeromonas salmonicida subsp. salmonicida Infection. Front Immunol. 2020, 10, 3011.