Wright Stain, pure, certified
Wright Stain, pure, certified
Wright Stain, pure, certified
Thermo Scientific Chemicals

Wright Stain, pure, certified

CAS: 68988-92-1 | C36H27Br4N3O5S+2 | 933.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 422641000
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
186.00
Save 27.35 (15%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
186.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS68988-92-1
IUPAC Name2,4,5,7-tetrabromo-9-(2-carboxyphenyl)-3,6-dihydroxy-10λ⁴-xanthen-10-ylium; 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium
Molecular FormulaC36H27Br4N3O5S
InChI KeyAXIKDPDWFVPGOD-UHFFFAOYSA-O
SMILESCN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C.OC(=O)C1=CC=CC=C1C1=C2C=C(Br)C(O)=C(Br)C2=[O+]C2=C(Br)C(O)=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Infrared spectrumConforms
Appearance (Form)Crystalline powder
UV spectrumComplies
BSC CertificationPasses
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only