Zinc oxide, ACS reagent
Zinc oxide, ACS reagent
Zinc oxide, ACS reagent
Thermo Scientific Chemicals

Zinc oxide, ACS reagent

CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 422740051
Price (EUR)
256,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
256,00
Each
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Complexometric>=99.0 %
Insoluble matter=<0.01 % (in dilute H2SO4)
Alkalinitypasses test
Sulfur compounds (as SO4)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only