Cesium chloride, 99+%, pure
Cesium chloride, 99+%, pure
Cesium chloride, 99+%, pure
Cesium chloride, 99+%, pure
Thermo Scientific Chemicals

Cesium chloride, 99+%, pure

CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 422851000
Price (EUR)
76,65
Online Exclusive
84,60
Save 7,95 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
76,65
Online Exclusive
84,60
Save 7,95 (9%)
Each
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White
Titration Argentometric99.0 to 101.0 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only