Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced
Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced
Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced
Thermo Scientific Chemicals

Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Catalog NumberQuantityPackaging
422981000100 gGlass bottle
Catalog number 422981000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Palladium (Pd)9.7 to 10.3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only