Petroleum ether, ACS reagent, boiling range 35-60°C
Petroleum ether, ACS reagent, boiling range 35-60°C
Petroleum ether, ACS reagent, boiling range 35-60°C
Thermo Scientific Chemicals

Petroleum ether, ACS reagent, boiling range 35-60°C

CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 422995000
Price (USD)
-
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Boiling point35°C to 60°C
Color scale=<10 APHA
AcidityPasses test
Appearance (Form)Clear liquid
Residue after evaporation=<0.001 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only