11-Bromoundecanoic Acid, 98%
11-Bromoundecanoic Acid, 98%
11-Bromoundecanoic Acid, 98%
Thermo Scientific Chemicals

11-Bromoundecanoic Acid, 98%

CAS: 2834-05-1 | C11H21BrO2 | 265.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 423160250
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.10
Save 6.45
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.10
Save 6.45
Each
Chemical Identifiers
CAS2834-05-1
IUPAC Name11-bromoundecanoic acid
Molecular FormulaC11H21BrO2
InChI KeyIUDGNRWYNOEIKF-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Crystalline chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only