Acetic acid, 99.7+%, ACS reagent
Acetic acid, 99.7+%, ACS reagent
Acetic acid, 99.7+%, ACS reagent
Thermo Scientific Chemicals

Acetic acid, 99.7+%, ACS reagent

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 423225000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS64-19-7
IUPAC Nameacetic acid
Molecular FormulaC2H4O2
InChI KeyQTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILESCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.7 %
Heavy metals (as Pb)=<0.5 ppm
Chloride (Cl)=<1 ppm
Sulfate (SO4)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only