Acetonitrile, 99.6%, ACS reagent
Acetonitrile, 99.6%, ACS reagent
Acetonitrile, 99.6%, ACS reagent
Thermo Scientific Chemicals

Acetonitrile, 99.6%, ACS reagent

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Catalog number 423250100
Price (USD)
559.65
Special offer
Online exclusive
Ends: 28-Dec-2026
658.00
Save 98.35
Each
Quantity:
10 L
Request bulk or custom format
Price (USD)
559.65
Special offer
Online exclusive
Ends: 28-Dec-2026
658.00
Save 98.35
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.5 %
Water=<0.3 % (K.F.)
Residue after evaporation=<0.005 %
Titrable acid=<0.008 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only