Acetic acid, ammonium salt, 97+%, ACS reagent
Acetic acid, ammonium salt, 97+%, ACS reagent
Acetic acid, ammonium salt, 97+%, ACS reagent
Thermo Scientific Chemicals

Acetic acid, ammonium salt, 97+%, ACS reagent

CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2 kg
Packaging:
Plastic bottle
Catalog number 423270020
Price (USD)
-
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<5 ppm
Appearance (Color)White
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Titration excess NaOH>=97.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only