Phthalic anhydride, ACS reagent
Phthalic anhydride, ACS reagent
Phthalic anhydride, ACS reagent
Thermo Scientific Chemicals

Phthalic anhydride, ACS reagent

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 423320020
Price (USD)
175.65
Special offer
Online exclusive
Ends: 28-Dec-2026
206.00
Save 30.35
Each
Quantity:
2 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
175.65
Special offer
Online exclusive
Ends: 28-Dec-2026
206.00
Save 30.35
Each
Chemical Identifiers
CAS85-44-9
IUPAC Name1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H4O3
InChI KeyLGRFSURHDFAFJT-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Sulfate (SO4)=<0.003 %
Appearance (Form)Flakes
Infrared spectrumConforms
Melting point129°C to 132°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only