Ammonium sulfate, 99+%, ACS reagent
Ammonium sulfate, 99+%, ACS reagent
Ammonium sulfate, 99+%, ACS reagent
Thermo Scientific Chemicals

Ammonium sulfate, 99+%, ACS reagent

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
数量:
500 g
5 kg
25 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 423400050
価格(JPY)
-
数量:
5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
さらに表示
Insoluble matter=<0.005 %
pH5.0 to 6.0 (5 % at 25.0°C)
Residue after ignition=<0.005 %
Chloride (Cl)=<5 ppm
Nitrate (NO3)=<0.001 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only