Ammonium sulfate, 99+%, ACS reagent
Ammonium sulfate, 99+%, ACS reagent
Ammonium sulfate, 99+%, ACS reagent
Thermo Scientific Chemicals

Ammonium sulfate, 99+%, ACS reagent

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
5 kg
25 kg
Packaging:
檢視全部
Plastic bottle
產品號碼 423405000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
檢視更多
規格Specification Sheet規格表
Insoluble matter=<0.005 %
pH5.0 to 6.0 (5 % at 25.0°C)
Residue after ignition=<0.005 %
Chloride (Cl)=<5 ppm
Nitrate (NO3)=<0.001 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only