Barium hydroxide octahydrate, 98+%, ACS reagent
Barium hydroxide octahydrate, 98+%, ACS reagent
Barium hydroxide octahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Barium hydroxide octahydrate, 98+%, ACS reagent

CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 423461000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12230-71-6
IUPAC Namebarium(2+) octahydrate dihydroxide
Molecular FormulaBaH18O10
InChI KeyZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals or flakes
Heavy metals (as Pb)=<5 ppm
Sulfide (S)(Limit about 0.001 %) Passes test
Insoluble matter=<0.01 % (in dilute HCl)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only