Cadmium chloride hemipentahydrate, ACS reagent
Cadmium chloride hemipentahydrate, ACS reagent
Cadmium chloride hemipentahydrate, ACS reagent
Thermo Scientific Chemicals

Cadmium chloride hemipentahydrate, ACS reagent

CAS: 7790-78-5 | 2·5 H2O | 228.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 423500025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7790-78-5
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals and/or chunks
Titration Complexometric79.5 to 81.0 % (CdCl2)
Nitrate and Nitrite (as NO3)=<0.003 %
Insoluble matter=<0.005 %
Sulfate (SO4)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only