Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.
Thermo Scientific Chemicals

Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
数量:
25 mL
500 mL
1 L
4 L
パッケージ:
Glass Bottle
製品番号(カタログ番号) 423550040
価格(JPY)
-
数量:
4 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
さらに表示
Color scale=<10 APHA
GC>=99.80 % (Excluding stabilizer)
Acetone and aldehydePasses test
Free chlorine(Cl) Passes test
Substance darkened by H2SO4Passes test
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only