Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Citric acid, ≥99.5%, ACS reagent, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
製品番号(カタログ番号) 423565000
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-92-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O7
InChI KeyKRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O
さらに表示
Appearance (Form)Crystalline powder
Titration with NaOH>=99.5 %
Carboniz. sub. by hot H2SO4(tartrates, etc) Passes test
Chloride (Cl)=<10 ppm
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only