Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, 98+%, ACS reagent

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
数量:
5 g
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 423580025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Sulfate (SO4)=<0.005 %
Calcium (Ca)=<0.005 %
Lead (Pb)=<0.002 %
Copper (Cu)=<0.002 %
Nickel (Ni)=<0.15 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only