Thermo Scientific Chemicals

Copper(II) sulfate pentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423610100
10 kg, Plastic Bucket, Each
Thermo Scientific Chemicals

Copper(II) sulfate pentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423610100
10 kg, Plastic Bucket, Each
Quantity
Packaging

Chemical Identifiers

CAS
7758-99-8
IUPAC Name
copper(2+) pentahydrate sulfate
Molecular Formula
CuH10O9S
InChI Key
JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
Appearance (Color)
Blue
Insoluble matter
=<0.005 %
Nitrogen compounds (as N)
=<0.002 %
Chloride (Cl)
=<10 ppm
Iron (Fe)
=<30 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text