Iron(III) nitrate nonahydrate, 98+%, ACS reagent
Iron(III) nitrate nonahydrate, 98+%, ACS reagent
Iron(III) nitrate nonahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, 98+%, ACS reagent

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
3 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 423710030
Price (USD)
270.65
Online Exclusive
301.00
Save 30.35 (10%)
Each
Quantity:
3 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
270.65
Online Exclusive
301.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS7782-61-8
IUPAC Nameiron(3+) nonahydrate trinitrate
Molecular FormulaFeH18N3O18
InChI KeySZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<50 ppm
Chloride (Cl)=<5 ppm
Sulfate (SO4)=<100 ppm
Appearance (Color)Almost white to light grey to light purple
Appearance (Form)Adhering crystals or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only