Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent
Thermo Scientific Chemicals

Ammonium iron(II) sulfate hexahydrate, 99%, ACS reagent

CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
3 kg
Packaging:
Plastic bottle
Catalog number 423720030
Price (USD)
543.65
Special offer
Online exclusive
Ends: 28-Dec-2026
639.00
Save 95.35
Each
Quantity:
3 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
543.65
Special offer
Online exclusive
Ends: 28-Dec-2026
639.00
Save 95.35
Each
Chemical Identifiers
CAS7783-85-9
IUPAC Nameλ²-iron(2+) diammonium hexahydrate disulfate
Molecular FormulaFeH20N2O14S2
InChI KeyMQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green
Zinc (Zn)=<0.003 %
Appearance (Form)Crystals
Titration with KMnO498.5 to 101.5 %
Magnesium (Mg)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only