Isopropanol, 99.6%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Isopropanol, 99.6%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Isopropanol, 99.6%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals

Isopropanol, 99.6%, ACS reagent, meets the requirements of Reag.Ph.Eur.

CAS: 67-63-0 | C3H8O | 60.096 g/mol
数量:
1 L
2.5 L
4 L
20 L
25 L
製品番号(カタログ番号) 423830200
価格(JPY)
-
数量:
20 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
さらに表示
Density(20/20°C) 0.784 to 0.787
Color scale=<10 APHA
GC>=99.5 %
Water=<0.2 % (K.F.)
Boiling point81°C to 83°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only