Lead(II) acetate trihydrate, 99+%, ACS reagent
Lead(II) acetate trihydrate, 99+%, ACS reagent
Lead(II) acetate trihydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, 99+%, ACS reagent

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423840025
Price (USD)
264.65
Special offer
Online exclusive
Ends: 28-Dec-2026
311.00
Save 46.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
264.65
Special offer
Online exclusive
Ends: 28-Dec-2026
311.00
Save 46.35
Each
Chemical Identifiers
CAS6080-56-4
IUPAC Nameλ²-lead(2+) diacetate trihydrate
Molecular FormulaC4H12O7Pb
InChI KeyMCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<10 ppm
Calcium (Ca)=<50 ppm
Potassium (K)=<50 ppm
Insoluble matter=<0.01 %
Nitrate and Nitrite (as NO3)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only