N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent
Acros Organics

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, C12H14N2.2ClH, CAS Number-1465-25-4 | CAS: 1465-25-4 | C12H16Cl2N2 | 259.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 423991000
HSN 29215990
Price (INR)
52,635.00
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
52,635.00
Each
Chemical Identifiers
CAS1465-25-4
IUPAC Namedihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Molecular FormulaC12H16Cl2N2
InChI KeyMZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to pink
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Titration Argentometric>=98.0 % (on dry substance)
Water=<5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only