Potassium oxalate monohydrate, 99%, ACS reagent
Potassium oxalate monohydrate, 99%, ACS reagent
Potassium oxalate monohydrate, 99%, ACS reagent
Acros Organics

Potassium oxalate monohydrate, 99%, ACS reagent

CAS: 6487-48-5 | C2H2K2O5 | 184.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 424025000
HSN 29171110
Price (INR)
8,953.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
8,953.00
Each
Chemical Identifiers
CAS6487-48-5
IUPAC Namedipotassium hydrate oxalate
Molecular FormulaC2H2K2O5
InChI KeyQCPTVXCMROGZOL-UHFFFAOYSA-L
SMILESO.[K+].[K+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with KMnO498.5 to 101.0 %
Chloride (Cl)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only