Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
Thermo Scientific Chemicals

Potassium hydrogen phthalate, ACS reagent, acidimetric standard

CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 424061000
Price (USD)
43.10
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
43.10
Each
Chemical Identifiers
CAS877-24-7
IUPAC Namepotassium 2-carboxybenzoate
Molecular FormulaC8H5KO4
InChI KeyIWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES[K+].OC(=O)C1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sodium (Na)=<0.005 %
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with NaOH99.95 to 100.05 % (on dried basis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only