Potassium ferricyanide, 99+%, ACS reagent
Potassium ferricyanide, 99+%, ACS reagent
Potassium ferricyanide, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium ferricyanide, 99+%, ACS reagent

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 424120050
Price (USD)
49.65
Online Exclusive
54.60
Save 4.95 (9%)
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.60
Save 4.95 (9%)
Each
Chemical Identifiers
CAS13746-66-2
IUPAC Nametripotassium hexacyanoirontris(ylium)
Molecular FormulaC6FeK3N6
InChI KeyQMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Ferro compounds=<0.05 % as ferrocyanide ion ¢Fe(CN)6!4-
Chloride (Cl)=<0.01 %
Sulfate (SO4)=<0.01 %
Appearance (Form)Crystals and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only