Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
数量:
500 g
2.5 kg
パッケージ:
Plastic Bottle
製品番号(カタログ番号) 424255000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
さらに表示
Loss on drying=<1.0 % (120°C)
Phosphate (PO4)=<0.001 %
Sulfate (SO4)=<0.003 %
Calcium (Ca)=<0.005 %
Magnesium (Mg)=<0.002 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only