Acetic acid, sodium salt trihydrate, 99+%, ACS reagent
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99+%, ACS reagent

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 424260025
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
133.00
Save 19.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
133.00
Save 19.35
Each
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
Heavy metals (as Pb)=<5 ppm
Potassium (K)=<0.005 %
Sulfate (SO4)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only