Sodium hydroxide, 97+%, ACS reagent, pellets
Sodium hydroxide, 97+%, ACS reagent, pellets
Sodium hydroxide, 97+%, ACS reagent, pellets
Thermo Scientific Chemicals

Sodium hydroxide, 97+%, ACS reagent, pellets

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
25 kg
Catalog number 424330250
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Pellets
Titration with HCl>=97.0 %
Heavy metals (as Ag)=<0.002 %
Sodium carbonate (Na2CO3)=<1.0 %
Nitrogen compounds (as N)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only