Sodium nitrite, 97+%, ACS reagent
Sodium nitrite, 97+%, ACS reagent
Sodium nitrite, 97+%, ACS reagent
Thermo Scientific Chemicals

Sodium nitrite, 97+%, ACS reagent

CAS: 7632-00-0 | NNaO2 | 69.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2 kg
Catalog number 424350050
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7632-00-0
IUPAC Namesodium nitrite
Molecular FormulaNNaO2
InChI KeyLPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES[Na+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with KMnO4>=97.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.01 %
Chloride (Cl)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only