Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent
Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent
Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Sodium phosphate, dibasic heptahydrate, 98+%, ACS reagent

CAS: 7782-85-6 | H15Na2O11P | 268.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 424385000
Price (USD)
97.10
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
97.10
Each
Chemical Identifiers
CAS7782-85-6
Molecular FormulaH15Na2O11P
InChI KeyLVXHNCUCBXIIPE-UHFFFAOYSA-L
SMILESO.[Na+].[Na+].OP([O-])([O-])=O
Molecular Weight (g/mol)268.06
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
pH8.7 to 9.3 (5 % solution at 25.0°C)
Chloride (Cl)=<10 ppm
Iron (Fe)=<10 ppm
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only