Sodium sulfite, 98+%, ACS reagent, anhydrous
Sodium sulfite, 98+%, ACS reagent, anhydrous
Sodium sulfite, 98+%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Sodium sulfite, 98+%, ACS reagent, anhydrous

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 424430010
Price (USD)
91.65
Special offer
Online exclusive
Ends: 28-Dec-2026
108.00
Save 16.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
91.65
Special offer
Online exclusive
Ends: 28-Dec-2026
108.00
Save 16.35
Each
Chemical Identifiers
CAS7757-83-7
IUPAC Namedisodium heptahydrate sulfite
Molecular FormulaH14Na2O10S
InChI KeyWMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Iodimetric>=98.0 %
Heavy metals (ICP-OES)=<0.001 %
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only