Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, 98+%, ACS reagent

CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 424480050
Price (USD)
57.30
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.30
Each
Chemical Identifiers
CAS10025-69-1
IUPAC Nameλ²-tin(2+) dihydrate dichloride
Molecular FormulaCl2H4O2Sn
InChI KeyFWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(in hydrochloric acid) Passes test
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder, crystals and chunks
Titration Iodimetric98.0 to 103.0 %
Sulfate (SO4)Passes test (limit about 0.003%)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only