Zinc sulfate heptahydrate, 99%, ACS reagent
Zinc sulfate heptahydrate, 99%, ACS reagent
Zinc sulfate heptahydrate, 99%, ACS reagent
Thermo Scientific Chemicals

Zinc sulfate heptahydrate, 99%, ACS reagent

CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
3 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 424600010
Price (USD)
157.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
157.00
Each
Chemical Identifiers
CAS7446-20-0
IUPAC Namezinc(2+) heptahydrate sulfate
Molecular FormulaH14O11SZn
InChI KeyRZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric99.0 to 103.0 %
Insoluble matter=<0.01 %
pH4.4 to 6.0 (5 % at 25°C)
Chloride (Cl)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only