Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.
Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.
Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.
Acros Organics

Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.

CAS: 79322-76-2 | C10H12O3 | 180.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 424750050
also known as 42475
HSN 29182910
Price (INR)
60,654.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
60,654.00
Each
Chemical Identifiers
CAS79322-76-2
IUPAC Namemethyl 4-[(1S)-1-hydroxyethyl]benzoate
Molecular FormulaC10H12O3
InChI KeyKAXLTAULVFFCNL-ZETCQYMHSA-N
SMILESCOC(=O)C1=CC=C(C=C1)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=88 %
Refractive index1.5300 to 1.5360 (20°C, 589 nm)
Appearance (Form)Viscous liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only