Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Thermo Scientific™

Nalgene™ Polypropylene Analytical Funnels

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호직경(미터법) 상단용량(미터법)제품 사이즈
4250-006563 mm64 mLCase of 36
4250-003535 mm10 mLCase of 36
4250-0035PK
4250-0035으로도 사용됨
35 mm10 mLPack of 12
4250-005554 mm41 mLCase of 36
4250-0055PK
4250-0055으로도 사용됨
54 mm41 mLPack of 12
4250-0065PK
4250-0065으로도 사용됨
63 mm64 mLPack of 12
4250-007576 mm114 mLCase of 36
4250-0075PK
4250-0075으로도 사용됨
76 mm114 mLPack of 6
4250-010098 mm254 mLCase of 24
4250-0100PK
4250-0100으로도 사용됨
98 mm254 mLPack of 4
4250-0160158 mm976 mLCase of 12
4250-0160PK
4250-0160으로도 사용됨
158 mm976 mLPack of 2
카탈로그 번호 4250-0065
제품 가격(KRW)
123,000
온라인 행사
Ends: 31-Dec-2026
144,000
할인액 21,000 (15%)
Case of 36
직경(미터법) 상단:
63 mm
용량(미터법):
64 mL
Unit Size:
Case of 36
제품 가격(KRW)
123,000
온라인 행사
Ends: 31-Dec-2026
144,000
할인액 21,000 (15%)
Case of 36

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable

Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.

사양
오토클레이브 가능Autoclavable
설명Funnel Analytical PP 65mm
직경(미터법) 스템7 mm
직경(미터법) 상단63 mm
길이(미터법) 스템58 mm
수량12/Pk.; 36/Cs.
용량(미터법)64 mL
물질Polypropylene
Unit SizeCase of 36