Nalgene™ Polypropylene Powder Funnels
Nalgene™ Polypropylene Powder Funnels
Nalgene™ Polypropylene Powder Funnels
Nalgene™ Polypropylene Powder Funnels
Thermo Scientific™

Nalgene™ Polypropylene Powder Funnels

Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopCapacity (Metric)Unit Size
4252-006564 mm68 mLCase of 36
4252-0065PK
also known as 4252-0065
64 mm68 mLPack of 12
4252-008079mm115 mLCase of 36
4252-0080PK
also known as 4252-0080
79mm115 mLPack of 12
4252-0100100 mm243 mLCase of 24
4252-0100PK
also known as 4252-0100
100 mm243 mLPack of 6
4252-0150145 mm716 mLCase of 24
4252-0150PK
also known as 4252-0150
145 mm716 mLPack of 4
Catalog number 4252-0065
Price (USD)
189.35
Online exclusive
242.76
Save 53.41
Case of 36
Diameter (Metric)Top:
64 mm
Capacity (Metric):
68 mL
Unit Size:
Case of 36
Price (USD)
189.35
Online exclusive
242.76
Save 53.41
Case of 36

Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.

  • Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
  • Suitable for general lab use, it is a durable replacement for glass funnels
  • External ribs prevent airlocks
  • Autoclavable

Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.

Specifications
AutoclavableAutoclavable
DescriptionFunnel Powder PP 65mm
Diameter (Metric) Stem17 mm
Diameter (Metric)Top64 mm
Length (Metric) Stem22mm
Quantity12/Pk.; 36/Cs.
Capacity (Metric)68 mL
Height (Metric)67 mm
MaterialPolypropylene
Unit SizeCase of 36